Gene Transcripts

Gene Symbol
DPP10
Entrez Gene ID
57628
Gene Name
dipeptidyl peptidase like 10
Gene Aliases
DPL2, DPPY, DPRP-3, DPRP3
Chromosome Location
Chr. 2: 114442358 - 115845752 on Build GRCh38
UniGene ID
Hs.580539
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004360.3 NP_001004360.2 4 407
NM_001178034.1 NP_001171505.1 4 334
NM_001178036.1 NP_001171507.1 5 811
NM_001178037.1 NP_001171508.1 4 377
NM_001321905.1 NP_001308834.1 5 837
NM_001321906.1 NP_001308835.1 5 453
NM_001321907.1 NP_001308836.1 4 722
NM_001321908.1 NP_001308837.1 4 696
NM_001321909.1 NP_001308838.1 3 311
NM_001321910.1 NP_001308839.1 5 780
NM_001321911.1 NP_001308840.1 4 779
NM_001321912.1 NP_001308841.1 5 811
NM_001321913.1 NP_001308842.1 3 607
NM_001321914.1 NP_001308843.1 4 337
NM_020868.4 NP_065919.2 4 722
XM_017004565.1 XP_016860054.1 5 881
XM_017004566.1 XP_016860055.1 3 326
GenBank mRNA AB040925.2 - 4 289
AK055256.1 - 6 895
AK289967.1 - 4 421
AK296490.1 - 4 409
AK315147.1 - 4 526
AY172661.3 - 4 301
BC030832.1 - 4 758 4811771
BI597037.1 - 4 364
DQ857321.1 - 4 289
DQ857322.1 - 4 313
DQ857323.1 - 4 280

View Full Product Details