Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001895.3 | NP_001886.1 | 13 | 1702 | |
NM_177559.2 | NP_808227.1 | 14 | 1819 | ||
NM_177560.2 | NP_808228.1 | 12 | 1492 | ||
GenBank mRNA | AK302583.1 | - | 12 | 1380 | |
BC050036.1 | - | 13 | 1591 | 5298655 | |
BC053532.1 | - | 12 | 1521 | 3908058 | |
BC071167.1 | - | 12 | 1433 | 5772615 | |
J02853.1 | - | 13 | 1579 | ||
M55265.1 | - | 13 | 1574 | ||
S53149.1 | - | 13 | 1579 |