Gene Transcripts

Gene Symbol
MYBBP1A
Entrez Gene ID
10514
Gene Name
MYB binding protein 1a
Gene Aliases
P160, PAP2
Chromosome Location
Chr. 17: 4538896 - 4555386 on Build GRCh38
UniGene ID
Hs.22824
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105538.1 NP_001099008.1 1 142
NM_014520.3 NP_055335.2 1 142
XM_011523616.2 XP_011521918.1 1 123
GenBank mRNA AF147709.1 - 1 105
AK301622.1 - 1 114
AK303150.1 - 1 110
AK307948.1 - 1 142
BC050546.1 - 1 85 5495483
DC346952.1 - 1 142

View Full Product Details