Gene Transcripts

Gene Symbol
SSH1
Entrez Gene ID
54434
Gene Name
slingshot protein phosphatase 1
Gene Aliases
SSH1L
Chromosome Location
Chr. 12: 108782690 - 108857606 on Build GRCh38
UniGene ID
Hs.199763
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161330.1 NP_001154802.1 3 240
NM_001161331.1 NP_001154803.1 2 332
NM_018984.3 NP_061857.3 3 240
XM_005268984.4 XP_005269041.1 3 951
XM_011538497.1 XP_011536799.1 2 332
XM_011538499.1 XP_011536801.1 2 332
XM_011538500.2 XP_011536802.1 1 29
XM_011538501.2 XP_011536803.1 1 29
XM_017019491.1 XP_016874980.1 2 1178
XM_017019492.1 XP_016874981.1 2 222
XM_017019493.1 XP_016874982.1 2 84
XM_017019494.1 XP_016874983.1 2 179
GenBank mRNA AB072355.1 - 3 153
AB072356.1 - 3 153
AK095421.1 - 2 333
BC062341.1 - 3 232 6376684
BP363810.1 - 3 240

View Full Product Details