Gene Transcripts
- Gene Symbol
- PPP2R2C
- Entrez Gene ID
- 5522
- Gene Name
- protein phosphatase 2 regulatory subunit Bgamma
- Gene Aliases
- B55-GAMMA, IMYPNO, IMYPNO1, PR52, PR55G
- Chromosome Location
- Chr. 4: 6320578 - 6563600 on Build GRCh38
- UniGene ID
- Hs.479069
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001206994.1 |
NP_001193923.1
|
3, 4 |
242 |
|
NM_001206995.1 |
NP_001193924.1
|
3, 4 |
346 |
|
NM_001206996.1 |
NP_001193925.1
|
3, 4 |
396 |
|
NM_020416.3 |
NP_065149.2
|
2, 3 |
534 |
|
NM_181876.2 |
NP_870991.1
|
2, 3 |
188 |
|
XM_011513495.1 |
XP_011511797.1
|
2, 3 |
283 |
|
XM_011513496.1 |
XP_011511798.1
|
2, 3 |
298 |
|
GenBank mRNA |
AB209183.1 |
- |
1, 2 |
3810 |
|
AF086924.1 |
- |
2, 3 |
218 |
|
AF145027.1 |
- |
2, 3 |
381 |
|
AK093115.1 |
- |
2, 3 |
188 |
|
AK296474.1 |
- |
3, 4 |
242 |
|
AK316003.1 |
- |
4, 5 |
356 |
|
BC032954.2 |
- |
3, 4 |
396 |
4823280 |
BM551976.1 |
- |
|
534 |
|
DC327075.1 |
- |
3, 4 |
351 |
|
View Full Product Details