Gene Transcripts

Gene Symbol
NAB1
Entrez Gene ID
4664
Gene Name
NGFI-A binding protein 1
Gene Aliases
Chromosome Location
Chr. 2: 190648860 - 190692766 on Build GRCh38
UniGene ID
Hs.107474
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321312.1 NP_001308241.1 4, 5 1323
NM_001321313.1 NP_001308242.1 4, 5 1109
NM_001321314.1 NP_001308243.1 3, 4 1032
NM_005966.3 NP_005957.2 6, 7 1585
XM_005246582.1 XP_005246639.1 5, 6 1258
XM_005246583.1 XP_005246640.1 5, 6 1372
XM_005246586.1 XP_005246643.1 4, 5 1065
XM_011511219.2 XP_011509521.1 5, 6 1227
XM_017004170.1 XP_016859659.1 5, 6 1268
XM_017004171.1 XP_016859660.1 4, 5 1438
XM_017004172.1 XP_016859661.1 4, 5 1210
XM_017004173.1 XP_016859662.1 5, 6 1293
XM_017004174.1 XP_016859663.1 4, 5 1109
XM_017004176.1 XP_016859665.1 4, 5 1177
GenBank mRNA AF045451.1 - 3, 4 1032
AK292446.1 - 4, 5 1123
AK313542.1 - 3, 4 1130
BC035724.1 - 6, 7 1474 5721972
U47007.1 - 3, 4 1032

View Full Product Details