Gene Transcripts
- Gene Symbol
- SUPT20H
- Entrez Gene ID
- 55578
- Gene Name
- SPT20 homolog, SAGA complex component
- Gene Aliases
- C13, C13orf19, FAM48A, FP757, P38IP, SPT20
- Chromosome Location
- Chr. 13: 37009312 - 37059714 on Build GRCh38
- UniGene ID
- Hs.435815
- Species
- Homo sapiens
View Full Product Details