Gene Transcripts
- Gene Symbol
- SUPT20H
- Entrez Gene ID
- 55578
- Gene Name
- SPT20 homolog, SAGA complex component
- Gene Aliases
- C13, C13orf19, FAM48A, FP757, P38IP, SPT20
- Chromosome Location
- Chr. 13: 37009312 - 37059714 on Build GRCh38
- UniGene ID
- Hs.435815
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001014286.2 |
NP_001014308.2
|
10, 11 |
962 |
|
NM_001278480.1 |
NP_001265409.1
|
10, 11 |
962 |
|
NM_001278481.1 |
NP_001265410.1
|
10, 11 |
965 |
|
NM_001278482.1 |
NP_001265411.1
|
10, 11 |
1103 |
|
NM_017569.3 |
NP_060039.1
|
10, 11 |
1044 |
|
XM_005266447.2 |
XP_005266504.1
|
10, 11 |
938 |
|
XM_005266449.3 |
XP_005266506.1
|
10, 11 |
899 |
|
XM_005266451.3 |
XP_005266508.1
|
10, 11 |
935 |
|
XM_005266454.3 |
XP_005266511.1
|
9, 10 |
897 |
|
XM_005266455.2 |
XP_005266512.1
|
10, 11 |
938 |
|
XM_005266456.3 |
XP_005266513.1
|
9, 10 |
970 |
|
XM_005266457.3 |
XP_005266514.1
|
9, 10 |
896 |
|
XM_005266458.3 |
XP_005266515.1
|
9, 10 |
967 |
|
XM_005266460.2 |
XP_005266517.1
|
10, 11 |
938 |
|
XM_005266461.3 |
XP_005266518.1
|
10, 11 |
938 |
|
XM_005266462.4 |
XP_005266519.1
|
10, 11 |
938 |
|
XM_005266464.3 |
XP_005266521.1
|
10, 11 |
938 |
|
XM_005266465.2 |
XP_005266522.1
|
10, 11 |
935 |
|
XM_005266467.3 |
XP_005266524.1
|
10, 11 |
938 |
|
XM_017020653.1 |
XP_016876142.1
|
10, 11 |
935 |
|
XM_017020654.1 |
XP_016876143.1
|
9, 10 |
897 |
|
XM_017020655.1 |
XP_016876144.1
|
9, 10 |
896 |
|
XM_017020656.1 |
XP_016876145.1
|
9, 10 |
965 |
|
XM_017020657.1 |
XP_016876146.1
|
10, 11 |
935 |
|
XM_017020658.1 |
XP_016876147.1
|
9, 10 |
917 |
|
XM_017020659.1 |
XP_016876148.1
|
9, 10 |
896 |
|
XM_017020660.1 |
XP_016876149.1
|
10, 11 |
935 |
|
XM_017020661.1 |
XP_016876150.1
|
9, 10 |
896 |
|
GenBank mRNA |
AF093250.1 |
- |
10, 11 |
910 |
|
AJ130894.1 |
- |
10, 11 |
1000 |
|
AK027554.1 |
- |
9, 10 |
783 |
|
AK225376.1 |
- |
10, 11 |
898 |
|
AK292376.1 |
- |
10, 11 |
1048 |
|
AK304227.1 |
- |
9, 10 |
901 |
|
BC001145.1 |
- |
11, 12 |
1088 |
3533877 |
BC030686.2 |
- |
10, 11 |
937 |
4826077 |
View Full Product Details