Gene Transcripts

Gene Symbol
ZFP1
Entrez Gene ID
162239
Gene Name
ZFP1 zinc finger protein
Gene Aliases
ZNF475
Chromosome Location
Chr. 16: 75126285 - 75172234 on Build GRCh38
UniGene ID
Hs.388813
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318469.1 NP_001305398.1 4 1780
NM_001318471.1 NP_001305400.1 5 1836
NM_001318472.1 NP_001305401.1 3 1703
NM_001318473.1 NP_001305402.1 3 1653
NM_001318474.1 NP_001305403.1 3 1634
NM_001318475.1 NP_001305404.1 4 1792
NM_001318476.1 NP_001305405.1 4 1778
NM_153688.3 NP_710155.2 4 1761
XM_011522921.2 XP_011521223.1 6 1821
XM_017022983.1 XP_016878472.1 4 1948
XM_017022984.1 XP_016878473.1 5 2075
XM_017022985.1 XP_016878474.1 5 2080
XM_017022986.1 XP_016878475.1 6 2168
XM_017022987.1 XP_016878476.1 4 1792
GenBank mRNA AK091562.1 - 1 988
AK094761.1 - 4 1698
BC033774.1 - 1 1124
BC064604.1 - 4 1639
BC113863.1 - 2 1452
BC114471.1 - 2 1452
BC127184.1 - 2 1452
BX647144.1 - 2 1416

View Full Product Details