Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005116.5 | NP_005107.4 | 4 | 548 | |
NM_203327.1 | NP_976072.1 | 4 | 566 | ||
XM_011529414.2 | XP_011527716.1 | 4 | 553 | ||
XM_011529415.2 | XP_011527717.1 | 3 | 374 | ||
XM_011529416.2 | XP_011527718.1 | 3 | 683 | ||
XM_011529417.2 | XP_011527719.1 | 3 | 430 | ||
XM_017028171.1 | XP_016883660.1 | 4 | 587 | ||
XM_017028172.1 | XP_016883661.1 | 4 | 591 | ||
XM_017028173.1 | XP_016883662.1 | 4 | 571 | ||
XM_017028174.1 | XP_016883663.1 | 4 | 466 | ||
XM_017028176.1 | XP_016883665.1 | 4 | 562 | ||
XM_017028177.1 | XP_016883666.1 | 4 | 549 | ||
XM_017028178.1 | XP_016883667.1 | 4 | 562 | ||
XM_017028179.1 | XP_016883668.1 | 4 | 549 | ||
XM_017028181.1 | XP_016883670.1 | 4 | 565 | ||
GenBank mRNA | AF058319.1 | - | 2 | 173 | |
AF092511.1 | - | 3 | 469 | ||
AF164142.1 | - | 4 | 566 | ||
AJ269478.1 | - | 2 | 173 | ||
AJ292318.1 | - | 3 | 404 | ||
AK296304.1 | - | 4 | 556 | ||
AK297253.1 | - | 4 | 570 | ||
AY380556.1 | - | 2 | 173 | ||
BC013112.2 | - | 4 | 537 | 3923057 | |
D87075.2 | - | 4 | 548 | ||
EF032501.1 | - | 2 | 173 |