Gene Transcripts

Gene Symbol
SLC22A9
Entrez Gene ID
114571
Gene Name
solute carrier family 22 member 9
Gene Aliases
HOAT4, OAT4, OAT7, UST3H, ust3
Chromosome Location
Chr. 11: 63368871 - 63410878 on Build GRCh38
UniGene ID
Hs.502772
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080866.2 NP_543142.2 6 1282
XM_017017159.1 XP_016872648.1 6 2200
XM_017017160.1 XP_016872649.1 5 859
GenBank mRNA AB062418.1 - 6 1115
AB074812.1 - 6 1263
AF440402.1 - 6 1250
AJ295270.1 - 6 1074
AK074246.1 - 6 1282
BC022379.1 - 5 1127
BC126288.1 - 6 1164 8992004

View Full Product Details