Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013334.3 | NP_037466.2 | 1 | 349 | |
NM_021971.2 | NP_068806.1 | 1 | 349 | ||
GenBank mRNA | AF135421.1 | - | 1 | 241 | |
AK024319.1 | - | 1 | 326 | ||
AK291700.1 | - | 1 | 324 | ||
BC001141.1 | - | 1 | 288 | 3503869 | |
BC008033.1 | - | 1 | 288 | 3355099 | |
DA411701.1 | - | 1 | 349 |