Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013334.3 | NP_037466.2 | 8 | 1402 | |
NM_021971.2 | NP_068806.1 | 9 | 1321 | ||
GenBank mRNA | AF135421.1 | - | 9 | 1213 | |
AK024319.1 | - | 8 | 1379 | ||
AK026582.1 | - | 1 | 100 | ||
AK291700.1 | - | 9 | 1296 | ||
BC001141.1 | - | 9 | 1260 | 3503869 | |
BC008033.1 | - | 9 | 1260 | 3355099 |