Gene Transcripts

Gene Symbol
KIF3C
Entrez Gene ID
3797
Gene Name
kinesin family member 3C
Gene Aliases
Chromosome Location
Chr. 2: 25926586 - 25982574 on Build GRCh38
UniGene ID
Hs.21611
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002254.6 NP_002245.4 1 881
XM_005264299.2 XP_005264356.2 1 881
GenBank mRNA AF035621.1 - 1 377
AK315324.1 - 1 791
BC092406.1 - 1 766 6504352
BC130423.1 - 1 452 40146454
BC132783.1 - 1 452 40146805
BC144223.1 - 1 452 9052743
BX571741.1 - 1 805

View Full Product Details