Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002254.6 | NP_002245.4 | 1 | 1040 | |
XM_005264299.2 | XP_005264356.2 | 1 | 1040 | ||
GenBank mRNA | AF018164.1 | - | 1 | 534 | |
AF035621.1 | - | 1 | 536 | ||
AK315324.1 | - | 1 | 950 | ||
BC042486.1 | - | 2 | 926 | 3866587 | |
BC092406.1 | - | 1 | 925 | 6504352 | |
BC130423.1 | - | 1 | 611 | 40146454 | |
BC132783.1 | - | 1 | 611 | 40146805 | |
BC144223.1 | - | 1 | 611 | 9052743 | |
BX571741.1 | - | 1 | 964 |