Gene Transcripts

Gene Symbol
SUOX
Entrez Gene ID
6821
Gene Name
sulfite oxidase
Gene Aliases
Chromosome Location
Chr. 12: 55996776 - 56005525 on Build GRCh38
UniGene ID
Hs.558403
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000456.2 NP_000447.2 6 1665
NM_001032386.1 NP_001027558.1 5 1539
NM_001032387.1 NP_001027559.1 4 1430
XM_005269112.1 XP_005269169.1 5 1521
XM_017019905.1 XP_016875394.1 7 2211
XM_017019906.1 XP_016875395.1 5 1499
XM_017019907.1 XP_016875396.1 6 2280
XM_017019908.1 XP_016875397.1 4 1440
GenBank mRNA AK312763.1 - 4 1404
BC065193.1 - 4 1461 5227360
HM013717.1 - 3 1348
L31573.1 - 3 2071

View Full Product Details