Gene Transcripts

Gene Symbol
PRDX6
Entrez Gene ID
9588
Gene Name
peroxiredoxin 6
Gene Aliases
1-Cys, AOP2, HEL-S-128m, NSGPx, PRX, aiPLA2, p29
Chromosome Location
Chr. 1: 173477347 - 173488807 on Build GRCh38
UniGene ID
Hs.120
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004905.2 NP_004896.1 5 1475
GenBank mRNA AJ844622.2 - 1 230
AK095940.1 - 1 1689
BC035857.1 - 5 1475 5767682
BC053550.1 - 5 1425 6013037
D14662.1 - 5 1467
EU794652.1 - 5 1475

View Full Product Details