Gene Transcripts

Gene Symbol
BPHL
Entrez Gene ID
670
Gene Name
biphenyl hydrolase-like (serine hydrolase)
Gene Aliases
BPH-RP, MCNAA, VACVASE
Chromosome Location
Chr. 6: 3118376 - 3153198 on Build GRCh38
UniGene ID
Hs.10136
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302777.1 NP_001289706.1 7 1346
NM_004332.3 NP_004323.2 6 1118
NR_026648.2 8 1414
NR_026649.2 7 1186
GenBank mRNA AJ617684.1 - 6 766
AK297518.1 - 7 870
AK314909.1 - 7 1000
AL832502.1 - 6 3113
BC037778.1 - 8 1419 4799692
BC106901.1 - 6 833 40031474
X57653.1 - 2 210
X81372.1 - 7 914

View Full Product Details