Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253723.1 | NP_001240652.1 | 4 | 575 | |
NM_001253724.1 | NP_001240653.1 | 4 | 578 | ||
NM_020134.3 | NP_064519.2 | 4 | 658 | ||
GenBank mRNA | AF157634.1 | - | 3 | 500 | |
AF264015.1 | - | 4 | 662 | ||
AJ251275.1 | - | 3 | 500 | ||
AL713706.1 | - | 2 | 200 | ||
BC002874.2 | - | 4 | 575 | 3940393 | |
BT006871.1 | - | 3 | 500 | ||
DA410946.1 | - | 4 | 578 |