Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039702.2 | NP_001034791.1 | 3 | 346 | |
NM_018324.2 | NP_060794.1 | 3 | 346 | ||
XM_017016376.1 | XP_016871865.1 | 3 | 310 | ||
GenBank mRNA | AK001844.1 | - | 3 | 296 | |
AK001968.1 | - | 3 | 279 | ||
BC050372.1 | - | 3 | 313 | 5742377 | |
BI830605.1 | - | 3 | 344 |