Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278192.1 | NP_001265121.1 | 15 | 1935 | |
NM_001278193.1 | NP_001265122.1 | 14 | 1869 | ||
NM_001278194.1 | NP_001265123.1 | 16 | 2102 | ||
NM_198334.2 | NP_938148.1 | 17 | 2211 | ||
NM_198335.3 | NP_938149.2 | 18 | 2277 | ||
XM_017017412.1 | XP_016872901.1 | 15 | 1945 | ||
XM_017017413.1 | XP_016872902.1 | 17 | 2327 | ||
XM_017017414.1 | XP_016872903.1 | 17 | 2158 | ||
GenBank mRNA | AF144074.1 | - | 18 | 2166 | |
AJ000332.1 | - | 17 | 2084 | ||
AK026548.1 | - | 6 | 615 | ||
AK026997.1 | - | 17 | 2101 | ||
AK295806.1 | - | 14 | 1757 | ||
AK299819.1 | - | 16 | 1985 | ||
AK302752.1 | - | 15 | 1841 | ||
AK314370.1 | - | 17 | 2100 | ||
BC017433.2 | - | 7 | 716 | ||
BC017435.2 | - | 7 | 716 | ||
BC034439.1 | - | 8 | 1087 | ||
BC065266.1 | - | 16 | 1978 | 6155944 | |
D42041.1 | - | 17 | 2082 | ||
EU794613.1 | - | 17 | 2117 |