Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301129.1 | NP_001288058.1 | 4 | 375 | |
NM_001301130.1 | NP_001288059.1 | 3 | 277 | ||
NM_001301131.1 | NP_001288060.1 | 3 | 277 | ||
NM_021974.4 | NP_068809.1 | 3 | 277 | ||
NR_125371.1 | 3 | 336 | |||
XM_017028832.1 | XP_016884321.1 | 3 | 243 | ||
GenBank mRNA | AK311128.1 | - | 3 | 300 | |
AK311831.1 | - | 3 | 241 | ||
BC003582.1 | - | 3 | 255 | 3546712 | |
BP195499.1 | - | 3 | 243 | ||
CR456546.1 | - | 3 | 167 | ||
CR457021.1 | - | 3 | 152 | ||
DA083933.1 | - | 4 | 339 | ||
DA203006.1 | - | 3 | 300 | ||
DA372532.1 | - | 3 | 277 | ||
DN989996.1 | - | 3 | 252 |