Gene Transcripts

Gene Symbol
UBE3A
Entrez Gene ID
7337
Gene Name
ubiquitin protein ligase E3A
Gene Aliases
ANCR, AS, E6-AP, EPVE6AP, HPVE6A
Chromosome Location
Chr. 15: 25337234 - 25439086 on Build GRCh38
UniGene ID
Hs.598862
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000462.3 NP_000453.2 6 917
NM_130838.1 NP_570853.1 2 132
NM_130839.2 NP_570854.1 5 852
XM_005268267.4 XP_005268324.1 7 1219
XM_005268268.4 XP_005268325.1 6 1106
XM_005268269.4 XP_005268326.1 7 950
XM_005268270.4 XP_005268327.1 6 907
XM_005268271.4 XP_005268328.1 3 726
XM_006720675.3 XP_006720738.1 7 1171
XM_006720676.3 XP_006720739.1 4 416
XM_011521994.2 XP_011520296.1 7 1108
XM_011521995.2 XP_011520297.1 7 782
XM_017022544.1 XP_016878033.1 8 1144
XM_017022545.1 XP_016878034.1 7 1103
XM_017022546.1 XP_016878035.1 7 1011
XM_017022547.1 XP_016878036.1 6 1043
XM_017022548.1 XP_016878037.1 6 1038
XM_017022549.1 XP_016878038.1 6 845
XM_017022550.1 XP_016878039.1 6 997
XM_017022551.1 XP_016878040.1 7 1048
XM_017022552.1 XP_016878041.1 7 738
XM_017022553.1 XP_016878042.1 7 1053
XM_017022554.1 XP_016878043.1 3 1057
XM_017022555.1 XP_016878044.1 5 826
XM_017022556.1 XP_016878045.1 6 975
XM_017022557.1 XP_016878046.1 5 910
XM_017022558.1 XP_016878047.1 6 1106
XM_017022559.1 XP_016878048.1 3 726
GenBank mRNA AB194232.1 - 4 228
AF273050.1 - 3 187
AI378869.1 - 2 140
AK056239.1 - 5 820
AK291405.1 - 3 278
AK292514.1 - 5 462
BC002582.2 - 5 805 3160225
BE349076.1 - 7 485
L07557.1 - 2 127
U84404.1 - 7 674
X98031.1 - 3 157
X98032.1 - 1 88
X98033.1 - 3 148

View Full Product Details