Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001098670.1 | NP_001092140.1 | 5 | 507 | |
NM_001098671.1 | NP_001092141.1 | 5 | 547 | ||
NM_001318398.1 | NP_001305327.1 | 5 | 467 | ||
NM_153819.1 | NP_722541.1 | 5 | 573 | ||
XM_005273707.3 | XP_005273764.3 | 5 | 898 | ||
XM_011544718.1 | XP_011543020.1 | 5 | 898 | ||
XM_011544720.1 | XP_011543022.1 | 5 | 978 | ||
XM_011544721.1 | XP_011543023.1 | 5 | 510 | ||
XM_011544722.1 | XP_011543024.1 | 5 | 563 | ||
XM_011544723.2 | XP_011543025.1 | 5 | 512 | ||
XM_011544725.2 | XP_011543027.1 | 3 | 262 | ||
XM_017017082.1 | XP_016872571.1 | 4 | 1548 | ||
XM_017017083.1 | XP_016872572.1 | 4 | 1548 | ||
XM_017017084.1 | XP_016872573.1 | 5 | 541 | ||
XM_017017085.1 | XP_016872574.1 | 2 | 331 | ||
GenBank mRNA | AF043722.1 | - | 6 | 679 | |
AF043723.1 | - | 5 | 573 | ||
AF081194.1 | - | 5 | 480 | ||
AK092882.1 | - | 4 | 1243 | ||
AK122694.1 | - | 5 | 431 | ||
AK124023.1 | - | 4 | 2043 | ||
AK314637.1 | - | 4 | 458 | ||
BC110306.1 | - | 5 | 481 | ||
BC117151.1 | - | 5 | 571 | 40125702 | |
DA529151.1 | - | 5 | 547 | ||
U78170.1 | - | 5 | 480 | ||
Y12336.1 | - | 5 | 573 |