Gene Transcripts

Gene Symbol
UBE2O
Entrez Gene ID
63893
Gene Name
ubiquitin conjugating enzyme E2 O
Gene Aliases
E2-230K
Chromosome Location
Chr. 17: 76389532 - 76453206 on Build GRCh38
UniGene ID
Hs.16130
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022066.3 NP_071349.3 18 3350
XM_005257567.1 XP_005257624.1 18 3337
XM_017024934.1 XP_016880423.1 18 3262
XM_017024935.1 XP_016880424.1 18 3259
GenBank mRNA AB051521.1 - 18 3350
AK022940.1 - 5 848
AK024657.1 - 3 349
AK025999.1 - 10 1829
AK225412.1 - 10 1829
AL832432.1 - 11 2275
BC004525.2 - 3 165
BC022237.1 - 5 868
BC025977.1 - 5 749
BC036820.1 - 13 2486
BC051868.2 - 10 1693 6162673

View Full Product Details