Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004218.3 | NP_004209.2 | 4 | 594 | |
GenBank mRNA | AF498947.1 | - | 4 | 498 | |
AK225036.1 | - | 4 | 512 | ||
AK225642.1 | - | 4 | 514 | ||
AK297498.1 | - | 4 | 522 | ||
AK310573.1 | - | 4 | 285 | ||
AK312994.1 | - | 4 | 548 | ||
BC110081.1 | - | 4 | 500 | 40035112 | |
BT019535.1 | - | 4 | 498 | ||
BT019536.1 | - | 4 | 498 | ||
BX647356.1 | - | 2596 | |||
CR536494.1 | - | 4 | 498 | ||
CR541691.1 | - | 4 | 498 | ||
CX787570.1 | - | 2 | 145 | ||
D38516.1 | - | 4 | 507 | ||
EF560724.1 | - | 4 | 577 | ||
X79780.1 | - | 4 | 504 |