Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000370.3 | NP_000361.1 | 5 | 1106 | |
XM_006716468.3 | XP_006716531.1 | 3 | 1228 | ||
GenBank mRNA | BC041784.1 | - | 5 | 1092 | |
BC058000.1 | - | 5 | 1110 | 4610804 | |
D49488.1 | - | 5 | 1092 |