Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001443.2 | NP_001434.1 | 2 | 253 | |
GenBank mRNA | BC022287.1 | - | 2 | 238 | |
BC032801.1 | - | 2 | 164 | 5190267 | |
BP263155.1 | - | 2 | 97 | ||
CR542090.1 | - | 2 | 139 | ||
M10050.1 | - | 2 | 181 | ||
M10617.1 | - | 2 | 154 |