Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001976.4 | NP_001967.3 | 5 | 434 | |
NM_053013.3 | NP_443739.3 | 5 | 392 | ||
XM_005256521.2 | XP_005256578.1 | 5 | 820 | ||
XM_011523729.1 | XP_011522031.1 | 5 | 410 | ||
XM_017024346.1 | XP_016879835.1 | 5 | 827 | ||
GenBank mRNA | AK300662.1 | - | 5 | 362 | |
AK309864.1 | - | 6 | 485 | ||
AU143784.1 | - | 5 | 431 | ||
BC017249.2 | - | 5 | 349 | 4902149 | |
BJ995304.1 | - | 5 | 408 | ||
DA082822.1 | - | 5 | 392 | ||
DA566866.1 | - | 5 | 390 | ||
X16504.1 | - | 4 | 296 | ||
X51957.1 | - | 4 | 308 |