Gene Transcripts
- Gene Symbol
- PSME2
- Entrez Gene ID
- 5721
- Gene Name
- proteasome activator subunit 2
- Gene Aliases
- PA28B, PA28beta, REGbeta
- Chromosome Location
- Chr. 14: 24143365 - 24147316 on Build GRCh38
- UniGene ID
- Hs.434081
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002818.2 |
NP_002809.2
|
2, 3 |
154 |
|
GenBank mRNA |
AK026580.1 |
- |
2, 3 |
175 |
|
AK225876.1 |
- |
2, 3 |
117 |
|
AY771595.1 |
- |
2, 3 |
103 |
|
BC004368.1 |
- |
2, 3 |
104 |
3640584 |
BC019885.1 |
- |
2, 3 |
104 |
4994572 |
BC072025.1 |
- |
2, 3 |
103 |
6270762 |
BX161498.1 |
- |
2, 3 |
729 |
|
CR541657.1 |
- |
2, 3 |
89 |
|
CR541743.1 |
- |
2, 3 |
89 |
|
D45248.1 |
- |
2, 3 |
154 |
|
View Full Product Details