Gene Transcripts

Gene Symbol
ANAPC10
Entrez Gene ID
10393
Gene Name
anaphase promoting complex subunit 10
Gene Aliases
APC10, DOC1
Chromosome Location
Chr. 4: 144967117 - 145098541 on Build GRCh38
UniGene ID
Hs.480876
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256706.1 NP_001243635.1 3 300
NM_001256707.1 NP_001243636.1 4 373
NM_001256708.1 NP_001243637.1 3 506
NM_001256709.1 NP_001243638.1 5 441
NM_001256710.1 NP_001243639.1 3 300
NM_001256711.1 NP_001243640.1 4 373
NM_001256712.1 NP_001243641.1 3 289
NM_001318367.1 NP_001305296.1 3 338
NM_014885.4 NP_055700.2 3 289
NR_046345.1 2 173
XM_011531524.2 XP_011529826.1 4 529
XM_011531525.2 XP_011529827.1 3 316
XM_011531526.2 XP_011529828.1 3 316
XM_011531527.2 XP_011529829.1 3 316
XM_011531529.2 XP_011529831.1 3 316
XM_011531530.2 XP_011529832.1 4 332
XM_011531531.2 XP_011529833.1 3 316
XM_011531532.2 XP_011529834.1 3 290
XM_017007638.1 XP_016863127.1 3 316
XM_017007639.1 XP_016863128.1 4 965
XM_017007640.1 XP_016863129.1 3 357
XM_017007641.1 XP_016863130.1 4 785
XM_017007642.1 XP_016863131.1 4 530
XM_017007643.1 XP_016863132.1 3 316
XM_017007644.1 XP_016863133.1 3 290
GenBank mRNA AB012109.1 - 3 287
AF132794.1 - 3 244
AL080090.1 - 4 347
AV757106.2 - 3 348
BC005217.1 - 3 266 3929766
BF693880.1 - 3 298
BG494966.1 - 5 403
BG530912.1 - 4 344
BI488888.1 - 2 171
BI913723.1 - 3 339
BU175327.1 - 3 270
BU567906.1 - 3 509
CN305215.1 - 2 168
CV023091.1 - 2 188

View Full Product Details