Gene Transcripts
- Gene Symbol
- PRRG4
- Entrez Gene ID
- 79056
- Gene Name
- proline rich and Gla domain 4
- Gene Aliases
- PRGP4, TMG4
- Chromosome Location
- Chr. 11: 32829756 - 32858123 on Build GRCh38
- UniGene ID
- Hs.471695
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024081.5 |
NP_076986.1
|
3, 4 |
535 |
|
XM_006718313.3 |
XP_006718376.3
|
3, 4 |
714 |
|
XM_006718314.3 |
XP_006718377.1
|
3, 4 |
385 |
|
XM_017018275.1 |
XP_016873764.1
|
3, 4 |
695 |
|
GenBank mRNA |
AF326351.1 |
- |
3, 4 |
383 |
|
BC010052.2 |
- |
3, 4 |
535 |
3841745 |
BC063393.1 |
- |
3, 4 |
527 |
5183337 |
KU178766.1 |
- |
2, 3 |
274 |
|
KU178767.1 |
- |
2, 3 |
216 |
|
KU178768.1 |
- |
2, 3 |
274 |
|
View Full Product Details