Gene Transcripts

Gene Symbol
NRBP2
Entrez Gene ID
340371
Gene Name
nuclear receptor binding protein 2
Gene Aliases
TRG16, pp9320
Chromosome Location
Chr. 8: 143829776 - 143840974 on Build GRCh38
UniGene ID
Hs.521926
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178564.3 NP_848659.2 18 3352
XM_017013380.1 XP_016868869.1 17 3443
XM_017013383.1 XP_016868872.1 17 3296
GenBank mRNA AF318376.1 - 8 2691
AK094017.1 - 2 2337
AL137662.1 - 16 3281
BC071605.1 - 17 3515

View Full Product Details