Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001191037.1 | NP_001177966.1 | 4 | 395 | |
NM_002807.3 | NP_002798.2 | 4 | 395 | ||
XM_017004517.1 | XP_016860006.1 | 4 | 395 | ||
GenBank mRNA | AK312528.1 | - | 4 | 352 | |
BC001053.2 | - | 4 | 352 | ||
BC014013.2 | - | 4 | 364 | ||
BC020223.1 | - | 4 | 342 | ||
BC032445.1 | - | 4 | 367 | ||
BC039845.1 | - | 4 | 318 | ||
BC047897.1 | - | 4 | 317 | ||
BC064398.1 | - | 4 | 366 | ||
BC073833.1 | - | 4 | 319 | ||
BC094720.1 | - | 4 | 381 | 4826340 | |
BC112344.1 | - | 4 | 356 | 40008454 | |
BC114434.1 | - | 4 | 356 | 40008444 | |
D44466.1 | - | 4 | 313 | ||
DA385817.1 | - | 4 | 395 |