Gene Transcripts

Gene Symbol
FLRT3
Entrez Gene ID
23767
Gene Name
fibronectin leucine rich transmembrane protein 3
Gene Aliases
HH21
Chromosome Location
Chr. 20: 14322985 - 14337847 on Build GRCh38
UniGene ID
Hs.41296
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013281.3 NP_037413.1 2 391
NM_198391.2 NP_938205.1 3 585
XM_005260682.4 XP_005260739.1 3 1103
XM_011529204.2 XP_011527506.1 4 1240
XM_011529205.2 XP_011527507.1 4 902
GenBank mRNA AB040902.1 - 2 279
AF169677.1 - 2 288
AK027297.1 - 2 302
AK027694.1 - 1 20
AK074883.1 - 2 332
AK074909.1 - 2 260
AL529481.3 - 3 541
AY358319.1 - 1 125
BC020870.1 - 2 298 4606852
CD654700.1 - 2 352
DA502792.1 - 2 391

View Full Product Details