Gene Transcripts

Gene Symbol
C11orf58
Entrez Gene ID
10944
Gene Name
chromosome 11 open reading frame 58
Gene Aliases
IMAGE145052, SMAP
Chromosome Location
Chr. 11: 16738601 - 16758354 on Build GRCh38
UniGene ID
Hs.368226
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014267.5 NP_055082.1 5 1566
XM_017017143.1 XP_016872632.1 5 3825
GenBank mRNA BC007103.1 - 5 1520

View Full Product Details