Gene Transcripts

Gene Symbol
RAB11FIP2
Entrez Gene ID
22841
Gene Name
RAB11 family interacting protein 2
Gene Aliases
Rab11-FIP2, nRip11
Chromosome Location
Chr. 10: 118004916 - 118046921 on Build GRCh38
UniGene ID
Hs.173656
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014904.2 NP_055719.1 5 2083
XM_005269629.3 XP_005269686.1 6 2461
GenBank mRNA AB023158.1 - 5 2083
AY037299.1 - 5 2083
BX647717.1 - 4 390

View Full Product Details