Gene Transcripts

Gene Symbol
RNLS
Entrez Gene ID
55328
Gene Name
renalase, FAD dependent amine oxidase
Gene Aliases
C10orf59, RENALASE
Chromosome Location
Chr. 10: 88176458 - 88583860 on Build GRCh38
UniGene ID
Hs.149849
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031709.2 NP_001026879.2 2 334
NM_018363.3 NP_060833.1 2 334
XM_005269946.1 XP_005270003.1 2 260
XM_005269947.1 XP_005270004.1 2 260
XM_005269949.4 XP_005270006.1 2 260
XM_011539924.2 XP_011538226.1 2 260
XM_011539927.2 XP_011538229.1 2 260
XM_017016380.1 XP_016871869.1 2 260
XM_017016381.1 XP_016871870.1 2 260
XM_017016383.1 XP_016871872.1 2 260
GenBank mRNA AK002080.1 - 2 261
BC005364.1 - 2 222 3828014
BP332262.1 - 2 334

View Full Product Details