Gene Transcripts

Gene Symbol
PBLD
Entrez Gene ID
64081
Gene Name
phenazine biosynthesis like protein domain containing
Gene Aliases
HEL-S-306, MAWBP, MAWDBP
Chromosome Location
Chr. 10: 68282660 - 68332958 on Build GRCh38
UniGene ID
Hs.198158
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033083.1 NP_001028255.1 5 510
NM_022129.3 NP_071412.2 5 510
XM_005270028.3 XP_005270085.1 5 544
XM_011540060.2 XP_011538362.1 5 544
XM_017016513.1 XP_016872002.1 5 503
XM_017016514.1 XP_016872003.1 5 544
GenBank mRNA AB049758.1 - 3 372
AI114748.1 - 2 45
AK027673.1 - 5 478
AK092826.1 - 3 243
AK312275.1 - 4 301
BC009738.1 - 5 492 3879159
CB121561.1 - 5 511
GQ472233.1 - 5 492

View Full Product Details