Gene Transcripts

Gene Symbol
CCDC30
Entrez Gene ID
728621
Gene Name
coiled-coil domain containing 30
Gene Aliases
PFD6L, PFDN6L
Chromosome Location
Chr. 1: 42521240 - 42654664 on Build GRCh38
UniGene ID
Hs.729640
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK024625.1 - 3 278
AK302075.1 - 1 45
AL832217.1 - 6 876
AY639646.1 - 2 359
DC397297.1 - 1 45

View Full Product Details