Gene Transcripts

Gene Symbol
PELI3
Entrez Gene ID
246330
Gene Name
pellino E3 ubiquitin protein ligase family member 3
Gene Aliases
Chromosome Location
Chr. 11: 66466327 - 66477337 on Build GRCh38
UniGene ID
Hs.656875
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098510.1 NP_001091980.1 7 2192
NM_001243135.1 NP_001230064.1 7 2031
NM_001243136.1 NP_001230065.1 6 2039
NM_145065.2 NP_659502.2 8 2264
XM_011544884.2 XP_011543186.1 8 2267
XM_017017465.1 XP_016872954.1 6 2016
GenBank mRNA AK074201.1 - 8 2202
AK075253.1 - 4 1704
AK094060.1 - 7 2031
AL834395.1 - 8 2211
BC025723.1 - 7 2113 5198999
BC127759.1 - 2 1350

View Full Product Details