Gene Transcripts

Gene Symbol
KDELC2
Entrez Gene ID
143888
Gene Name
KDEL motif containing 2
Gene Aliases
Chromosome Location
Chr. 11: 108472106 - 108498432 on Build GRCh38
UniGene ID
Hs.83286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153705.4 NP_714916.3 2 315
XM_011542621.2 XP_011540923.1 2 288
XM_011542623.2 XP_011540925.1 2 282
GenBank mRNA AF533708.1 - 2 274
AK125519.1 - 1 97
AK313534.1 - 2 177
AY358616.1 - 1 384
BC036526.2 - 2 269 5266665
DN999478.1 - 2 323

View Full Product Details