Gene Transcripts

Gene Symbol
KDELC2
Entrez Gene ID
143888
Gene Name
KDEL motif containing 2
Gene Aliases
Chromosome Location
Chr. 11: 108472106 - 108498432 on Build GRCh38
UniGene ID
Hs.83286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153705.4 NP_714916.3 8 2964
XM_011542621.2 XP_011540923.1 8 2937
XM_011542623.2 XP_011540925.1 6 2517
GenBank mRNA AF533708.1 - 8 2923
AK098206.1 - 4 1930
AK123743.1 - 1 813
AK125519.1 - 8 2864
AY358616.1 - 7 3033
BC036526.2 - 8 2918 5266665
BX648867.1 - 2 1589

View Full Product Details