Gene Transcripts
- Gene Symbol
- HIGD1A
- Entrez Gene ID
- 25994
- Gene Name
- HIG1 hypoxia inducible domain family member 1A
- Gene Aliases
- HIG1, RCF1a
- Chromosome Location
- Chr. 3: 42782908 - 42804535 on Build GRCh38
- UniGene ID
- Hs.593134
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001099668.1 |
NP_001093138.1
|
3, 4 |
521 |
|
NM_001099669.1 |
NP_001093139.1
|
3, 4 |
374 |
|
NM_014056.3 |
NP_054775.2
|
3, 4 |
350 |
|
GenBank mRNA |
AF077034.1 |
- |
3, 4 |
302 |
|
AF145385.1 |
- |
3, 4 |
289 |
|
AK225461.1 |
- |
3, 4 |
274 |
|
AK225571.1 |
- |
3, 4 |
305 |
|
AK311815.1 |
- |
3, 4 |
305 |
|
AL110233.1 |
- |
3, 4 |
320 |
|
BC000601.1 |
- |
3, 4 |
288 |
3346355 |
BC009583.1 |
- |
3, 4 |
281 |
4093421 |
BC009594.1 |
- |
3, 4 |
287 |
4109190 |
BC070277.1 |
- |
3, 4 |
303 |
4686431 |
BG708126.1 |
- |
3, 4 |
355 |
|
BP208784.1 |
- |
3, 4 |
520 |
|
BP370118.1 |
- |
3, 4 |
296 |
|
CR457385.1 |
- |
2, 3 |
228 |
|
View Full Product Details