Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318934.1 | NP_001305863.1 | 2 | 419 | |
NM_001318935.1 | NP_001305864.1 | 2 | 419 | ||
NM_003071.3 | NP_003062.2 | 2 | 419 | ||
NM_139048.2 | NP_620636.1 | 2 | 419 | ||
XM_011513091.1 | XP_011511393.1 | 2 | 419 | ||
XM_011513092.1 | XP_011511394.1 | 2 | 419 | ||
XM_011513093.1 | XP_011511395.1 | 2 | 419 | ||
XM_017007078.1 | XP_016862567.1 | 2 | 398 | ||
XM_017007079.1 | XP_016862568.1 | 2 | 419 | ||
GenBank mRNA | AB209052.1 | - | 2 | 201 | |
AK291231.1 | - | 2 | 366 | ||
BC005260.1 | - | 2 | 383 | ||
BC015498.1 | - | 2 | 375 | ||
BC030976.1 | - | 2 | 419 | ||
BC044659.1 | - | 2 | 387 | 6015181 | |
L34673.1 | - | 2 | 378 | ||
S64671.1 | - | 1 | 90 |