Gene Transcripts

Gene Symbol
NEPRO
Entrez Gene ID
25871
Gene Name
nucleolus and neural progenitor protein
Gene Aliases
C3orf17, NET17
Chromosome Location
Chr. 3: 113002444 - 113019733 on Build GRCh38
UniGene ID
Hs.591288
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319109.1 NP_001306038.1 4 449
NM_001319110.1 NP_001306039.1 3 294
NM_001319112.1 NP_001306041.1 5 462
NM_001319114.1 NP_001306043.1 5 508
NM_001319115.1 NP_001306044.1 5 504
NM_015412.3 NP_056227.2 6 634
NR_134969.1 4 449
XM_005247276.2 XP_005247333.1 6 601
XM_017006102.1 XP_016861591.1 6 597
XM_017006103.1 XP_016861592.1 5 492
GenBank mRNA AK024325.1 - 6 592
AK293218.1 - 4 387
AK295165.1 - 5 393
AK295954.1 - 3 231
AK301161.1 - 5 422
AK304083.1 - 5 437
AL117573.1 - 6 576
AL537045.3 - 5 429
BC014673.2 - 5 1092
BC067743.1 - 6 594 4812681
CR749341.1 - 4 450

View Full Product Details