Gene Transcripts
- Gene Symbol
- SSUH2
- Entrez Gene ID
- 51066
- Gene Name
- ssu-2 homolog (C. elegans)
- Gene Aliases
- C3orf32, SSU-2, fls485
- Chromosome Location
- Chr. 3: 8619400 - 8745035 on Build GRCh38
- UniGene ID
- Hs.736935
- Species
- Homo sapiens
View Full Product Details