Gene Transcripts

Gene Symbol
SSUH2
Entrez Gene ID
51066
Gene Name
ssu-2 homolog (C. elegans)
Gene Aliases
C3orf32, SSU-2, fls485
Chromosome Location
Chr. 3: 8619400 - 8745035 on Build GRCh38
UniGene ID
Hs.736935
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256748.1 NP_001243677.1 12 1122
NM_001256749.1 NP_001243678.1 12 1143
NM_015931.2 NP_057015.1 12 1289
NR_046358.1 13 1370
XM_011533774.1 XP_011532076.1 20 2838
XM_017006510.1 XP_016861999.1 13 5603
XM_017006511.1 XP_016862000.1 19 2526
XM_017006512.1 XP_016862001.1 19 2695
XM_017006513.1 XP_016862002.1 18 2531
XM_017006514.1 XP_016862003.1 18 2437
XM_017006515.1 XP_016862004.1 20 3592
XM_017006516.1 XP_016862005.1 15 1953
XM_017006517.1 XP_016862006.1 15 1906
XM_017006518.1 XP_016862007.1 20 2817
XM_017006519.1 XP_016862008.1 13 5548
XM_017006520.1 XP_016862009.1 13 1899
XM_017006521.1 XP_016862010.1 16 2057
XM_017006522.1 XP_016862011.1 16 1948
XM_017006523.1 XP_016862012.1 14 1651
XM_017006524.1 XP_016862013.1 15 1889
XM_017006525.1 XP_016862014.1 18 2458
XM_017006526.1 XP_016862015.1 13 1537
XM_017006527.1 XP_016862016.1 19 2580
XM_017006528.1 XP_016862017.1 13 1576
XM_017006529.1 XP_016862018.1 13 1606
XM_017006530.1 XP_016862019.1 15 2039
XM_017006531.1 XP_016862020.1 14 1961
XM_017006533.1 XP_016862022.1 19 2613
XM_017006534.1 XP_016862023.1 19 2559
XM_017006538.1 XP_016862027.1 17 2156
XM_017006539.1 XP_016862028.1 12 1268
XM_017006540.1 XP_016862029.1 13 1447
XM_017006541.1 XP_016862030.1 10 1089
XM_017006542.1 XP_016862031.1 10 1067
GenBank mRNA AB024705.1 - 12 1262
AK057972.1 - 13 1366
AK093025.1 - 1 1831
AK093066.1 - 18 2319
AK122938.1 - 17 2887
AK225925.1 - 13 1366
AK300175.1 - 12 1122
BC052614.1 - 12 1126 6300057

View Full Product Details