Gene Transcripts

Gene Symbol
POGLUT1
Entrez Gene ID
56983
Gene Name
protein O-glucosyltransferase 1
Gene Aliases
C3orf9, CLP46, KDELCL1, KTELC1, MDS010, MDSRP, Rumi, hCLP46
Chromosome Location
Chr. 3: 119468938 - 119494708 on Build GRCh38
UniGene ID
Hs.231750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152305.2 NP_689518.1 3 281
NR_024265.1 3 281
XM_006713705.3 XP_006713768.1 2 92
XM_017006878.1 XP_016862367.1 2 112
XM_017006879.1 XP_016862368.1 3 227
XM_017006880.1 XP_016862369.1 3 227
GenBank mRNA AF168711.1 - 3 232
AK075444.1 - 3 217
AK222937.1 - 3 241
AK295984.1 - 3 256
AK301002.1 - 3 256
AK315367.1 - 3 256
AY298903.1 - 3 267
AY358581.1 - 3 224
BC030614.2 - 3 267 4796951
BC048810.1 - 3 225 5243487
BP347179.1 - 3 281
CD517010.1 - 3 197

View Full Product Details