Gene Transcripts
- Gene Symbol
- ATG9A
- Entrez Gene ID
- 79065
- Gene Name
- autophagy related 9A
- Gene Aliases
- APG9L1, MGD3208, mATG9
- Chromosome Location
- Chr. 2: 219219380 - 219229688 on Build GRCh38
- UniGene ID
- Hs.323363
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX537984.1 |
- |
2, 3 |
218 |
|
View Full Product Details